Cystine dihydrochloride, weißgraues Puder

Lieferant: MP Biomedicals

Synonyme: DL-Cystine dihydrochloride

0210145205
ICNA0210145205EA 67 EUR
ICNA0210145205
Cystine dihydrochloride, weißgraues Puder
Cystine dihydrochloride

Bitte beachten Research Use Only (RUO).

Formel: C₆H₁₄Cl₂N₂O₄S₂
Lagertemperatur: Raumtemperatur
CAS-Nummer: 90350-38-2

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
FTIR Conforms to Standard

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR