L(-)-Cystin ~99%, Crystalline

Lieferant: MP Biomedicals

Synonyme: (R)-(-)-Cystin, (2R,2'R)-(-)-3,3'-Dithiobis(2-aminopropansäure), (H-Cys-OH)2

0210145425 0210145480 0210145490 0210145491
ICNA0210145425EA 36.1 EUR
ICNA0210145425 ICNA0210145480 ICNA0210145490 ICNA0210145491
L(-)-Cystin ~99%, Crystalline
L(-)-Cystin

50 mg/ml, 1 M HCl.


Solubility: 50 mg/ml, 1 M HCl.


Bitte beachten Research Use Only (RUO).

Formel: HOOCCH(NH₂)CH₂SSCH₂CH(NH₂)COOH
Molecular Weight: 240.3 g/mol
Siedepunkt: 428
Schmelzpunkt: >240 °C (Zers., Lit.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00064228
CAS-Nummer: 56-89-3
Merck Index: 13,02811

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
Purity 98.5 - 101.0%
FTIR Conforms to Standard
Moisture ≤0.5%
Optical Rotation (c=1,1N HCl) –210 to –230°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR