Desoxycholsäure freie Säure ≥99%, weißes Pulver

Lieferant: MP Biomedicals

Synonyme: 3α,12α-Dihydroxycholansäure, 12α-Hydroxylithocholsäure, DCA, Deoxycholsäure, 3α,12α-Dihydroxy-5β-cholan-24-säure, 3α12α-Dihydroxycholan-24-säure, 12alpha-Hydroxylithocholsäure, 3α,12α-Dihydroxy-5β-cholansäure

0210149610 0210149680
ICNA0210149610EA 27.3 EUR
ICNA0210149610 ICNA0210149680
Desoxycholsäure freie Säure ≥99%, weißes Pulver
Desoxycholsäure

Deoxycholic acid is a bile acid formed by metabolism of cholate.


Deoxycholic acid has been used in a modified procedure to recover 40 to 80% of a protein from a 1 μg/ml solution. It forms complexes with fatty acid.


The free acid is soluble at 0,24 g/L in water at 15 °C.


Bitte beachten Research Use Only (RUO).

Formel: C₂₄H₄₀O₄
Molecular Weight: 392.58 g/mol
Siedepunkt: 504
Schmelzpunkt: 190
Lagertemperatur: Raumtemperatur
MDL: MFCD00003673
CAS-Nummer: 83-44-3
Merck Index: 13,02916

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
Purity ≥99%
FTIR Conforms to Standard
Melting Point 168 - 178 °C
Extinction Coefficient EmM = 9,4 (Lit.)
Optical Rotation +54,1° (c=2 in MeOH) (Lit.)
pka 6,58 (Lit.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR