(2,4-Dichlorphenoxy)essigsäure, bräunliches Pulver

Lieferant: MP Biomedicals
Gefahr

0210155380
ICNA0210155380EA 57.4 EUR
ICNA0210155380
(2,4-Dichlorphenoxy)essigsäure, bräunliches Pulver
(2,4-Dichlorphenoxy)essigsäure

2,4-Dichlorophenoxyacetic acid is a herbicide.


2,4-Dichlorophenoxyacetic acid is a herbicide.


Soluble in ethanol (100 mg/ml - clear to hazy, faint yellow solution), methanol (50 mg/ml - clear, colourless solution).

Formel: (C₆H₃Cl₂)OCH₂COOH
Siedepunkt: 346 °C (1013 hPa)
Schmelzpunkt: 134…140 °C
Dichte: 1,563 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00004300
CAS-Nummer: 94-75-7
EINECS: 202-361-1
UN: 3077
ADR: 6.1,III
Merck Index: 13,02825

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR