2,2′-Bipyridin ≥99,5%, Pulver
Lieferant: MP Biomedicals
|
Gefahr
|
Synonyme:
α,α'-Dipyridyl, 2,2'-Bipyridin
α,α'-Dipyridyl is a bidentate ligand employed in transition metal catalysis and aluminum initiated polymerisation (inorganic syntheses).
Reagent for iron determinations.
2,2′-Bipyridyl has been used in the preparation of copper(II)-bipyridine-naringenin complex. It may be used in the preparation of 2,2′-bipyridyl hydrobromide. Ligand for copper in greener oxidation of alcohols under aerobic conditions.
Soluble in ethyl alcohol (50 mg/ ml - clear solution).
Bitte beachten Research Use Only (RUO).
Formel:
C₁₀H₈N₂ Molecular Weight: 156,19 g/mol Schmelzpunkt: 72 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006212 CAS-Nummer: 366-18-7 UN: 2811 ADR: 6.1,III |
Spezifikation Testergebnisse
Identity Test | Passes |
Purity | ≥99.5% |
FTIR | Conforms to Standard |
Melting Point | 65 - 75 °C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...