Äsculinhydrat ≥97,5%, weißes Pulver

Lieferant: MP Biomedicals

Synonyme: 6,7-Dihydroxycumarin-6β-D-glucopyranosid, 6-O-(-D-Glucosyl)aesculetin, Schillerstoff, Aesculin

0210165505
ICNA0210165505EA 56.1 EUR
ICNA0210165505
Äsculinhydrat ≥97,5%, weißes Pulver
Äsculinhydrat

Esculin Monohydrate protects cells against DNA damage and reduces ROS levels. It is also a carcinogenesis inhibitor and displays vitamin P activity.


Esculin is a coumarin glycoside that demonstrates antioxidant activitites and protection against chemically-induced carcinogenesis. Esculin is utilized in microbiology for the isolation and identification of Enterococcus (group D streptococci).


Soluble in water (0,1% aq soln) Clear and Colorless within 15 minutes, hot alcohol, methanol, pyridine, ethyl acetate, acetic acid.


Bitte beachten Research Use Only (RUO).

Formel: C₁₅H₁₆O₉·xH₂O
Molecular Weight: 340,29 g/mol (anhy)
Lagertemperatur: Raumtemperatur
MDL: MFCD00149492
CAS-Nummer: 531-75-9

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
Purity ≥97.5%
FTIR Conforms to Standard
UV/Vis: λmax (ethanol) 334 - 338 nm
pH (0,1% aq soln) 4,6 - 5,2
Loss on Drying ≤8%
Optical Rotation -84 ° to -87 °(c=2, 50% Dioxane)
Solubility (0.1% aq soln) Clear and colourless within 15 minute

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR