2-(4-Iodphenyl)-3-(4-nitrophenyl)-5-phenyltetrazoliumchlorid ≥99%, gelbes Pulver

Lieferant: MP Biomedicals
Achtung

0210206625 0210206605 02102066.1 02102066.5 0210206601
ICNA0210206625EA 1803 EUR
ICNA0210206625 ICNA0210206605 ICNA02102066.1 ICNA02102066.5 ICNA0210206601
2-(4-Iodphenyl)-3-(4-nitrophenyl)-5-phenyltetrazoliumchlorid ≥99%, gelbes Pulver
2-(4-Iodophenyl)-3-(4-nitrophenyl)-5-phenyltetrazolium chloride
Formel: C₁₉H₁₃ClIN₅O₂
Schmelzpunkt: 227…229 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00011961
CAS-Nummer: 146-68-9
EINECS: 205-676-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR