4-Hydroxy-2-chinolincarbonsäure, Pulver
Lieferant: MP Biomedicals
|
Achtung
|
Synonyme:
Kynurensäure, 4-Hydroxychinaldinsäure, KYNA
02102124.2
ICNA02102124.2EA
52.3
EUR
ICNA02102124.2
4-Hydroxy-2-chinolincarbonsäure, Pulver
4-Hydroxy-2-chinolincarbonsäure
Kynurenic acid is a non-selective antagonist at NMDA and AMPA/kainate receptors.
- Blocks kainic acid neurotoxicity
- Blocks nicotinic acetycholine receptors
Soluble in water, hot alcohol and 1,0 M NaOH (50 mg/ml - clear, pale yellow solution); insoluble in ether.
Bitte beachten Research Use Only (RUO).
Formel:
C₁₀H₇NO₃ Molecular Weight: 189,17 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149476 CAS-Nummer: 492-27-3 |
Spezifikation Testergebnisse
Identity Test | Passes |
FTIR | Conforms to Standard |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...