Lecithin (aus Sojabohne), dunkelgelbes Pulver

Lieferant: MP Biomedicals

0210214780 0210214783 0210214790 0210214791
ICNA0210214780EA 34.1 EUR
ICNA0210214780 ICNA0210214783 ICNA0210214790 ICNA0210214791
Lecithin (aus Sojabohne), dunkelgelbes Pulver
Lecithin

Lecithin from Soy, Refined, is a natural mixture of phospholipids consisting of phosphatidyl choline, phosphatidyl ethanolamine, and phosphatidyl inositol. In soybean lecithin the predominant fatty acid is polyunsaturated linoleic acid, amounting to about 70% of the fatty acids present.
Lecithin is used for preparation of vesicle suspensions commonly called liposomes or as monolayers.
A major structural phospholipid in brain, comprising approx. 15% of total lipid; primarily localized to gray matter.


  • Presentation: Dark Yellow Powder
  • Storage temperature: Store at -0 °C.


Storage temperature: -20 °C

Lagertemperatur: Gefrierschrank
MDL: MFCD00147406
CAS-Nummer: 8002-43-5

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
Source Soybean (refined oil removed)
Moisture ≤2%
Acetone Insolubles ≥95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR