D(-)-Mannitol ≥96%, Pulver

Lieferant: MP Biomedicals

Synonyme: (2R,3R,4R,5R)-(-)-1,2,3,4,5,6-Hexanhexol, D(-)-Mannit

0210224801 02102248.5 0210224805
ICNA0210224801EA 122 EUR
ICNA0210224801 ICNA02102248.5 ICNA0210224805
D(-)-Mannitol ≥96%, Pulver
D(-)-Mannitol

Mannitol is carbohydrate found in plants and plant exudates.


A sugar alcohol sweet tastant. Used in sweetness inhibition studies.


A sugar alcohol sweet tastant. Used in sweetness inhibition studies.


Soluble in water (1 g/5,5 ml), alcohol (1 g/83 ml), glycerol (1 g/18 ml); more soluble in hot water; soluble in pyridine, aniline, and aqueous solutions of alkalies; insoluble in ether.

Formel: HOH₂C(CH(OH))₄CH₂OH
Molecular Weight: 182.17 g/mol
Siedepunkt: 290…295 °C (4 hPa)
Schmelzpunkt: 163…172 °C
Dichte: 1.49 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00064287
CAS-Nummer: 69-65-8
EINECS: 200-711-8
Merck Index: 13,05769
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=163f556dca45086e05ff490ddc8eeb65d04aea05&vid=408e3f5566e309064ce68988cdd160305c5b8c0a&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR