Ölsäure, Liquid, Practical

Lieferant: MP Biomedicals

Synonyme: cis-9-Octadecensäure

0210250090 0210250091
ICNA0210250090EA 92 EUR
ICNA0210250090 ICNA0210250091
Ölsäure, Liquid, Practical
Ölsäure

Oleic acid is a mono-unsaturated omega-9 fatty acid created by dehydrogenation of stearic acid. It is then further dehydrogenated and elongated to create members of the n-9 family of fatty acids.


Oleic acid is used as emollient. Small amounts of oleic acid are used as an excipient in pharmaceuticals. It is used as an emulsifying or solubilizing agent in aerosol products.


Oleic Acid activates protein kinase C in hepatocytes. It uncouples oxidative phosphorylati It inhibits 2,4-dinitrophenol -stimulated ATPase. Action reversed by adding serum albumin.


Soluble in methanol (40 mg/ml - clear solution), chloroform, benzene or fixed and volatile oils. Insoluble in water.


Bitte beachten Research Use Only (RUO).

Formel: C₁₈H₃₄O₂
Molecular Weight: 282.47 g/mol
Siedepunkt: 286
Schmelzpunkt: 16.7
Flammpunkt: 180
Lagertemperatur: Gefrierschrank
MDL: MFCD00064242
CAS-Nummer: 112-80-1
Merck Index: 13,06898
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e5aecc5dce4d0318386a45d7adb7c5822631010a&vid=892a2df4ecab3679b06983b84cd27ad334ef961f&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR