Olivenöl, Reagenzqualität

Lieferant: MP Biomedicals

0210250290
ICNA0210250290EA 80.3 EUR
ICNA0210250290
Olivenöl, Reagenzqualität
Olive oil

Olive oil is commonly used in cooking, cosmetics, pharmaceuticals, and soaps and as a fuel for traditional oil lamps.


The health benefits of olive oil result from its higher proportion of monounsaturated fats, as well as its antioxidant constituents: hydroxytyrosol, oleuropein and oleocanthal.


The health benefits of olive oil result from its higher proportion of monounsaturated fats, as well as its antioxidant constituents: hydroxytyrosol, oleuropein and oleocanthal.


Very slightly soluble in methanol, n-octanol; insoluble in cold water.


Bitte beachten Research Use Only (RUO).

Lagertemperatur: Raumtemperatur
MDL: MFCD00131764
CAS-Nummer: 8001-25-0

Bestellen

Spezifikation Testergebnisse

Identity Test Passes

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR