4-Aminobenzoesäure, weißes Pulver

Lieferant: MP Biomedicals

Synonyme: 4-Carbonsäureanilin, PABA

0210256980
ICNA0210256980EA 80.2 EUR
ICNA0210256980
4-Aminobenzoesäure, weißes Pulver
4-Aminobenzoesäure

p-Aminobenzoic acid is an organic compound used in the treatment of fibrotic skin disorders like Peyronie's disease. It is employed as an intermediate in the synthesis of folate (Vitamin BX), azo dyes and cross linking agents.


4-Aminobenzoic acid acts as a gastric acid secretion inhibitor.


It is also useful in biomedical sector for the preparation of local anesthetic and ointments. It is widely used in industries to make pharmaceutical products, flavors and preservatives. It is actively involved as a UV-blocking ingredient in sun tan cosmetics.


Soluble in water (1 g/170 ml), alcohol, ether, ethyl acetate, glacial acetic acid, warm glycerine, alkali hydroxides and carbonates; slightly soluble in benzene, chloroform.

Formel: C₇H₇NO₂
Molecular Weight: 137,14 g/mol
Dichte: 1,374 g/mL (25 °C) (Lit.)
MDL: MFCD00007894
CAS-Nummer: 150-13-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR