Orthoperiodsäure ≥99%, blassgelbes kristallines Pulver

Lieferant: MP Biomedicals
Gefahr

Synonyme: Überiodsäure, ortho-Periodsäure

0210259580
ICNA0210259580EA 84.7 EUR
ICNA0210259580
Orthoperiodsäure ≥99%, blassgelbes kristallines Pulver
Orthoperiodsäure

Periodic acid is a compound used to stain the cytoplasm of cells found in the tubular pattern, mucus, and apical surfaces of cells of various lumen and glycoproteins on SDS-PAGE gels.


Periodic acid is used for staining of cell cytoplasm . Also periodic acid can be used in oxidation reactions converting sulphides into sulphones, catalysed by chromium(VI) oxide and oxidation reactions converting alcohols to aldehydes and ketones. Periodic acid can also react with the 1,2 glycol linkage of carbohydrates forming aldehydes in tissue sections which can be colored with Schiff bases, thus allowing for the staining of these tissues. Furthermore, studies show that periodic acid is not effective in staining cancer cells in solid nests, which are better stained using Grimelius .


Freely soluble in water (100 mg/ml), alcohol, nitric acid; slightly soluble in ether.

Formel: H₅IO₆
Molecular Weight: 227.94 g/mol
Schmelzpunkt: 122
MDL: MFCD00011344
CAS-Nummer: 10450-60-9
UN: 3085
ADR: 5.1 II
Merck Index: 13,07248
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=39e1882cdf619581f467ea2429265a700b5205d5&vid=27e789fa0f30c08d62d5a8d0afc3acd8f6469c12&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Form Yellowish Crystalline Powder
Storage Store at Room Temperature (15 - 30 °C). Store Desiccated.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR