L-α-Phenylalanin ≥99%, weißes Pulver

Lieferant: MP Biomedicals

Synonyme: H-Phe-OH, H-L-Phe-OH, L-α-Phenylalanin, 3-Phenyl-L(-)-alanin, 3-Phenyl-L-alanin, (S)-(-)-2-Amino-3-phenylpropansäure

0210262383 0210262390 0210262391 0210262325 0210262380
ICNA0210262383EA 225 EUR
ICNA0210262383 ICNA0210262390 ICNA0210262391 ICNA0210262325 ICNA0210262380
L-α-Phenylalanin ≥99%, weißes Pulver
L-α-Phenylalanin

L-Phenylalanine is one of the 20 amino acids. It is an essential amino acid. It contains an aromatic side chain, which contributes to ultraviolet absorbance and fluorescence properties.


L-Phenylalanine is used in the manufacture of food and drink products and sold as a nutritional supplement for its reputed analgesic and antidepressant effects. It is a direct precursor to the neuromodulator phenylethylamine, a commonly used dietary supplement.


L-Phenylalanine is used in the manufacture of food and drink products and sold as a nutritional supplement for its reputed analgesic and antidepressant effects. It is a direct precursor to the neuromodulator phenylethylamine, a commonly used dietary supplement.


Soluble in water, hydrochloric acid (50 mg/ml-clear, colorless solution), aqueous (20 mg/ml-clear, colorless solution; powder may need to be ground slightly with a mortar and pestle before solubilization); very slighly soluble in methanol and ethanol.

Formel: C₉H₁₁NO₂
Siedepunkt: 313 °C (1013 hPa)
Schmelzpunkt: 270…275 °C (Lit.)
Dichte: 0,754 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00064227
CAS-Nummer: 63-91-2
EINECS: 200-568-1
Merck Index: 14,07271

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
Purity ≥99%
FTIR Conforms to Standard
UV/VIS: l max (water) 257 ± 3 nm 257.3 nm
Optical Rotation (c=2, water) -35° ± 4°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR