L(+)-Rhamnose Monohydrat ∼99%, Pulver

Lieferant: MP Biomedicals

Synonyme: Isodulcit, 6-Desoxy-L(+)-mannose Monohydrat

0210280905 0210280910 0210280925 0210280950 0210280980
ICNA0210280905EA 56.5 EUR
ICNA0210280905 ICNA0210280910 ICNA0210280925 ICNA0210280950 ICNA0210280980
L(+)-Rhamnose Monohydrat ∼99%, Pulver
L(+)-Rhamnose Monohydrat

Soluble in water (100 mg/ml - clear, colorless solution).


Soluble in water (100 mg/ml - clear, colorless solution)


Bitte beachten Research Use Only (RUO).

Formel: C₆H₁₂O₅·1H₂O
Molecular Weight: 182.17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00149363
CAS-Nummer: 10030-85-0
Merck Index: 12,08338

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
Purity ~99%
FTIR Conforms to Standard
Melting Point 91 - 101 °C
Moisture Report Result
Optical Rotation (c=4, H₂O, NH₃) +8.2 ± 1°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR