D(−)-Salicin, Pulver

Lieferant: MP Biomedicals

0210283305 0210283380 0210283310 0210283325
ICNA0210283305EA 61.7 EUR
ICNA0210283305 ICNA0210283380 ICNA0210283310 ICNA0210283325
D(−)-Salicin, Pulver
D(−)-Salicin

Salicin is an alcoholic β-glucoside derived from willow bark that produces anti-inflammatory (including analgesic and antipyretic) effects very similar to that of aspirin Salicin is a substrate for β-glucosidase.


Salicin is used as an analgesic and antipyretic. It is also used as a substrate for beta-glucosidase and an inhibitor of Cox. It finds application as an anti-inflammatory agent. It is involved in antipyretic action without causing gastric injury.


Biochem/physiol Actions: It acts as a non-selective COX inhibitor, exhibiting IC50 values > 100 µM for COX-1 and COX-2.


Freely soluble in water (50 mg/ml-clear,colorless solution); soluble in alkalies, pyridine, glacial acetic acid; practically insoluble in ether and chloroform.

Formel: C₁₃H₁₈O₇
Schmelzpunkt: 199 - 201 °C
Dichte: 1,42 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00006590
CAS-Nummer: 138-52-3
EINECS: 205-331-6
Merck Index: 12,08476

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
FTIR Conforms to Standard
194 - 204 °C Melting Point
Optical Rotation −65° ± 5° (c=3, water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR