Natriumchlorid 99,0-100,5%, Pulver USP

Lieferant: MP Biomedicals

Synonyme: Saline, NaCl, Sea salt, Halite

02102892.5 0210289201 0210289205 0210289210
ICNA02102892.5EA 56.6 EUR
ICNA02102892.5 ICNA0210289201 ICNA0210289205 ICNA0210289210
Natriumchlorid 99,0-100,5%, Pulver USP
Natriumchlorid

Soluble in water (357 mg/ml at 25 °C; 384 mg/ml at 100 °C), glycerol (100 mg/ml); very slightly soluble in ethanol. Solubility in water is decreased by HCl; almost insoluble in concentrated HCl. Aqueous solutions are at a neutral pH (6,7 to 7,3). Aqueous solutions of sodium chloride are stable at room temperature.


Sodium chloride is used in a wide variety of biochemical applications, including intravenous fluids (normal saline is 0,9% w/v in water), density gradients, a diluent to increase ionic strength in buffers or culture media and in salt-out procedures in the isolation of DNA. It has also been used in high concentrations for preservation of foods since bacteria cannot grow in high salt conditions. A salt-ice mixture in the ratio of approximately 33 g sodium chloride to 100 g ice (at -1 °C) will drop in temperature to as low as -21 °C, depending on the rate of stirring and the size of the ice chunks.

Formel: NaCl
Molecular Weight: 58.44 g/mol
Siedepunkt: 1413
Schmelzpunkt: 801
MDL: MFCD00003477
CAS-Nummer: 7647-14-5
Merck Index: 14,08599

Bestellen

Spezifikation Testergebnisse

Synonyms Salt; Dendritis; Halite; Purex; Rock salt; saline; sterling; table salt; top flake
Form Powder
Storage Store at Room Temperature (15 - 30 °C)
Heavy Metals ≤5 ppm
Loss on Drying ≤0,5%
Sterilisation of Solutions It can be sterilised by autoclaving or sterile filtering
Aluminum ≤0,2 μg/g
Magnesium and Calcium ≤0,01%
Arsenic ≤1 μg/g
Iron ≤2 μg/g
Limit of Phosphates ≤0,0025%
Limit of Potassium ≤0,05%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR