Dinatriumsuccinat Hexahydrat ≥98%, weißes kristallines Pulver

Lieferant: MP Biomedicals
Achtung

Synonyme: Natriumsuccinat dibasisch Hexahydrat, 1,4-Butandisäure Dinatriumsalz Hexahydrat, Bernsteinsäure Dinatriumsalz Hexahydrat

0210292980 0210292990 0210292991
ICNA0210292980EA 29.6 EUR
ICNA0210292980 ICNA0210292990 ICNA0210292991
Dinatriumsuccinat Hexahydrat ≥98%, weißes kristallines Pulver
Dinatriumsuccinat Hexahydrat

Soluble in water (100 mg/ml - clear, colorless solution).


Use as a TCA (Krebs cycle) supplement in cell culture applications. Sodium succinate dibasic hexahydrate can be used to study x-ray crystallography, protein structural analysis, proteomics and optimization reagents. Sodium succinate dibasic hexahydrate has been used to determine the calorimetric and diffractometric evidence for the sequential crystallization of buffer components and the consequential pH swing in frozen solutions. Sodium succinate dibasic hexahydrate has also been used to provide insights into the mechanism of Pseudomonas dacunhae aspartate ß-decarboxylase from rapid-scanning stopped-flow kinetics.


Soluble in water (100 mg/ml - clear, colorless solution).

Formel: NaOOCCH₂CH₂COONa·6H₂O
Molecular Weight: 270.14 g/mol
Schmelzpunkt: 300
Lagertemperatur: Raumtemperatur
MDL: MFCD00149117
CAS-Nummer: 6106-21-4
Merck Index: 13,08754

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
Purity ≥98%
FTIR Conforms to Standard
pH (1% aq soln) 7.7 - 8.7
Moisture Content 35 - 40%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR