Sorbinsäure ≥99%, gelbes kristallines Pulver

Lieferant: MP Biomedicals
Achtung

0210293780
ICNA0210293780EA 29.9 EUR
ICNA0210293780
Sorbinsäure ≥99%, gelbes kristallines Pulver
Sorbinsäure

Sorbic acid (2,4-hexadienoic acid) is a straight chain unsaturated fatty acid. It is an antimicrobial agent. The antimicrobial action of sorbic acid is due to its inhibitory influence on various enzymes in the microbial cell such as enzymes involved in carbohydrate metabolism such as enolase and lactate dehydrogenase, enzymes of citric acid cycles.


Sorbic acid is an antimicrobial agents often used as preservatives in food and drinks to prevent the growth of mold, yeast, and fungi.
Sorbic acid can be used to inhibit bacterial, yeast and fungal sulfhydryl enzymes by inhibiting amino acid uptake.


Sorbic acid can be used to inhibit bacterial, yeast and fungal sulfhydryl enzymes by inhibiting amino acid uptake.


Moderately soluble in water.


Bitte beachten Research Use Only (RUO).

Formel: CH₃CH=CHCH=CHCOOH
Siedepunkt: >190 °C (1013 hPa) (Lit.)
Schmelzpunkt: 130…140 °C
Dichte: 1,2 g/cm³ (20 °C) (Lit.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00002703
CAS-Nummer: 110-44-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=440c50b4fc52d9ff0b52afa7ba596c860f95e0e7&vid=f9b0ff409010208781696e9ca3314c02aaf1af36&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
Purity ≥99%
FTIR Conforms to Standard
Melting Point 130 - 140 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR