Sulfanilamid, weißes Pulver

Lieferant: MP Biomedicals

Synonyme: Anilin-4-sulfonsäureamid, 4-Aminobenzolsulfonamid, 4-Aminobenzolsulfonsäureamid

0210298890
ICNA0210298890EA 159 EUR
ICNA0210298890
Sulfanilamid, weißes Pulver
Sulfanilamid

Sulfanilamide is used used as Antibacterial agent. It is also used to treat Vaginal infection caused by fungus.


Sulfonamide antibiotic that blocks the synthesis of dihydrofolic acid by inhibiting the enzyme dihydropteroate synthase. Mode of Action: A competitive inhibitor of dihydropteroate synthestase to block the synthesis of folic acid. Anti-microbial Spectrum: Gram positive, Gram negative, Chlamydia. Mode of Resistance: Alteration of dihydropteroate synthase or alternative pathway for folic acid synthesis.


Sulphonamide antibiotic that blocks the synthesis of dihydrofolic acid by inhibiting the enzyme dihydropteroate synthase. Mode of action: a competitive inhibitor of dihydropteroate synthestase to block the synthesis of folic acid. Anti-microbial spectrum: gram positive, gram negative, chlamydia. Mode of resistance: alteration of dihydropteroate synthase or alternative pathway for folic acid synthesis.


Soluble in 1 g dissolves in 37 ml alcohol, 5 ml acetone; glycerol, ether, benzene, petroleum ether.


Bitte beachten Research Use Only (RUO).

Formel: H₂NC₆H₄SO₂NH₂
Molecular Weight: 172,21 g/mol
Schmelzpunkt: 160…170 °C
Dichte: 1,08 g/mL (Lit.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00007939
CAS-Nummer: 63-74-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a76bb9ab6bdf23e9d846b24d4f57047dea1d5071&vid=f2f1c5e47a127cd33d3b4a2a31b1057c375456eb&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
FTIR Conforms to Standard
Melting Point 160 - 170°C
UV/VIS: λmax (water) 255 - 259 nm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR