Thiamin Hydrochlorid, Pulver

Lieferant: MP Biomedicals

Synonyme: Thiaminiumchlorid, Antiberiberifaktor Hydrochlorid, antineuritisches Vitamin Hydrochlorid, Aneurin Hydrochlorid, Thiaminchlorid Hydrochlorid, Vitamin B1 Hydrochlorid

0210302825 0210302880 0210302883 0210302891
ICNA0210302825EA 31.1 EUR
ICNA0210302825 ICNA0210302880 ICNA0210302883 ICNA0210302891
Thiamin Hydrochlorid, Pulver
Thiamin Hydrochlorid

Thiamine (Vitamin B1) is one of the essential vitamins and is required for carbohydrate metabolism.


Thiamine hydrochloride is used as a food-additive to add a brothy/meaty flavor to gravies or soups. It is also used as food supplement.


Soluble in HCl (50 mg/ml - clear, colorless solution).


Bitte beachten Research Use Only (RUO).

Formel: C₁₂H₁₈Cl₂N₄OS
Lagertemperatur: Raumtemperatur
MDL: MFCD00012780
CAS-Nummer: 67-03-8

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
FTIR Conforms to Standard
pH (1% aq soln) 2,7 - 3,4
OD400 (1% aq soln) ≤0.25
Loss on Drying ≤5,0%
Residue on Ignition ≤0,2%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR