p-Toluolsulfonsäure Monohydrat ≥99%, weißes Pulver

Lieferant: MP Biomedicals
Achtung

Synonyme: Toluol-4-sulfonsäure Monohydrat, 4-Methylbenzolsulfonsäure Monohydrat

0210308710
ICNA0210308710EA 42 EUR
ICNA0210308710
p-Toluolsulfonsäure Monohydrat ≥99%, weißes Pulver
p-Toluolsulfonsäure Monohydrat

p-Toluenesulfonic acid monohydrate is an organic-soluble acid catalyst in organic synthesis. The catalyst has been incorporated in the esterification of carboxylic acids or transesterification of esters.


p-Toluenesulfonic acid monohydrate has been used as a reducing agent for the reductive amination of ketones and aldehydes. In the presence of p-Toluenesulfonic acid monohydrate novel deazaflavin-cholestane hybrid compounds have been synthesized in a condensation reaction. 2-Phenylethyl alpha-glucoside has also been synthesized in the presence of p-Toluenesulfonic acid monohydrate.


Soluble in water (50 mg/ml-clear, colorless solution) or ethanol.


Bitte beachten Research Use Only (RUO).

Formel: C₇H₈O₃S·1H₂O
Molecular Weight: 190,22 g/mol
Schmelzpunkt: 97,..107 °C
Dichte: 1,23...1.24 g/cm³ (20 °C) (Lit.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00142137
CAS-Nummer: 6192-52-5
UN: 2585
ADR: 8,III

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
Purity >99%
FTIR Conforms to Standard
Melting Point 97 - 107°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR