D(+)-Tryptophan ≥99%, Pulver
Lieferant: MP Biomedicals
0210314425
0210314480
0210314401
ICNA0210314425EA
172
EUR
ICNA0210314425
ICNA0210314480
ICNA0210314401
D(+)-Tryptophan ≥99%, Pulver
D(+)-Tryptophan
D-Tryptophan is an amino acid and a unnatural isomer of tryptophan.
Soluble in water or hot alcohol. Also soluble in 0,5 M Hydrochloric Acid (50 mg plus 1 ml 0,5 M HCl) giving clear to slightly hazy solution.
Formel:
C₁₁H₁₂N₂O₂ Schmelzpunkt: 280…283 °C |
MDL:
MFCD00005647 CAS-Nummer: 153-94-6 EINECS: 205-819-9 Merck Index: 14,09797 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...