D(+)-Tryptophan ≥99%, Pulver

Lieferant: MP Biomedicals

0210314425 0210314480 0210314401
ICNA0210314425EA 172 EUR
ICNA0210314425 ICNA0210314480 ICNA0210314401
D(+)-Tryptophan ≥99%, Pulver
D(+)-Tryptophan

D-Tryptophan is an amino acid and a unnatural isomer of tryptophan.


Soluble in water or hot alcohol. Also soluble in 0,5 M Hydrochloric Acid (50 mg plus 1 ml 0,5 M HCl) giving clear to slightly hazy solution.

Formel: C₁₁H₁₂N₂O₂
Schmelzpunkt: 280…283 °C
MDL: MFCD00005647
CAS-Nummer: 153-94-6
EINECS: 205-819-9
Merck Index: 14,09797

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR