Polysorbat 20, Flüssigkeit

Lieferant: MP Biomedicals

0210316880 0210316890 0210316891
ICNA0210316880EA 31.2 EUR
ICNA0210316880 ICNA0210316890 ICNA0210316891
Polysorbat 20, Flüssigkeit
Polysorbat 20

Polysorbate 20 is nonionic surfactant derived from sorbitan ester.


  • Refractive Index: n20/D 1.468(Lit.)
  • Viscosity: 400 cps at 25 °C.(Lit.)
  • Critical Micelle Concentration (CMC): 0.06mM


Polysorbate 20 is typically used as an emulsifying agent for the preparation of stable oil-in-water emulsions, particularly in pharmaceutical applications. It has been used in pre-extraction of membranes to remove peripheral proteins. It has been used as a blocking agent on nitrocellulose.


Bitte beachten Research Use Only (RUO).

Formel: C₅₈H₁₁₄O₂₆
Siedepunkt: >100 °C (Lit.)
Dichte: 1,095 g/mL (Lit.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00165986
CAS-Nummer: 9005-64-5

Bestellen

Spezifikation Testergebnisse

Identity Test A Passes
Identity Test B Passes
FTIR Conforms to Reference
Solubility (1g/20mL) Clear, Foamy Solution
Specific Gravity 1.08-1.113
Water Content ≤3%
Saponification Number 40-50


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR