Kaffeesäure ≥98%, gelbes Pulver

Lieferant: MP Biomedicals

Synonyme: 3,4-Dihydroxyzimtsäure, 3-(3,4-Dihydroxyphenyl)propensäure, 3,4-Dihydroxycinnamsäure

0210479701 0210479705 0210479725
ICNA0210479701EA 38.3 EUR
ICNA0210479701 ICNA0210479705 ICNA0210479725
Kaffeesäure ≥98%, gelbes Pulver
Kaffeesäure

Caffeic acid is used as an anti-oxidant.


Inhibits the synthesis of leukotrienes that are involved in immunoregulation, inflammation and allergy. Inhibits Cu2+-induced LDL oxidation.


Soluble in ethanol (50 mg/ml) (Lit.).

Formel: C₉H₈O₄
Molecular Weight: 180.16 g/mol
Siedepunkt: 366
MDL: MFCD00004392
CAS-Nummer: 331-39-5
Merck Index: 12,01673

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
Purity ≥98%
FTIR Conforms to Standard


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR