Peptone, powder, bacteriological grade, MP Biomedicals
Lieferant: MP Biomedicals
0210480801
0210480805
ICNA0210480801EA
228
EUR
ICNA0210480801
ICNA0210480805
Peptone, powder, bacteriological grade, MP Biomedicals
Medien für die Mikrobiologie
Basismaterialien für die Mikrobiologie
Soluble in water (100 mg/ml - clear to slightly hazy solution).
- Aerobic Plate Count ≤1000 cfu/g
- Yeasts/Molds <100 cfu/g; Nitrogen ≥14%
Peptone is used in culture media in bacteriology, the growth requirements of most non fastidious organisms will be fulfilled by the range of amino acids, peptides and proteoses in this mixture.