5-Brom-4-chlor-3-indolylphosphat Dinatriumsalz (BCIP-Na) ≥99%, weißgraues Puder

Lieferant: MP Biomedicals

Synonyme: 5-Bromo-4-chloro-1H-indol-3-yl phosphate sodium, X-phosphate disodium salt, Sodium 5-bromo-4-chloro-1H-indol-3-yl phosphate

0215004283 0215004291 0215004250 0215004280
ICNA0215004283EA 186 EUR
ICNA0215004283 ICNA0215004291 ICNA0215004250 ICNA0215004280
5-Brom-4-chlor-3-indolylphosphat Dinatriumsalz (BCIP-Na) ≥99%, weißgraues Puder
5-Brom-4-chlor-3-indolylphosphat Dinatriumsalz (BCIP-Na)

A chromogenic substrate for alkaline phosphatase.


For the colorimetric detection of alkaline phosphatase-labeled molecules, 5-Bromo-4-chloro-3-indolyl phosphate (BCIP) and Nitro Blue Tetrazolium (NBT) are available. The BCIP/NBT substrate system is versatile and functions in a variety of applications, including Northern Southern, and Western blotting, in situ hybridization, and immunohistochemistry. BCIP disodium salt is soluble in water. It may be used to prepare a stock solution, which in combination with NBT and a reaction buffer, form a substrate solution for alkaline phosphatase. This substrate system, when incubated with alkaline phosphatase, produces an insoluble NBT diformazan product that is easily observable with its purple color.


Disodium salt is soluble in water (50 mg/ml); insoluble in dimethylformamide.

Formel: C₈H₄BrClNNa₂O₄P
Molecular Weight: 370.43 g/mol
Schmelzpunkt: >300 °C (Lit.)
Lagertemperatur: Gefrierschrank
MDL: MFCD00036757
CAS-Nummer: 102185-33-1

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
Purity ≥99%
FTIR Conforms to Reference
Loss on Drying ≤10%
UV/VIS: I max (water) 291 ± 3nm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR