Mineralöl, Flüssigkeit

Lieferant: MP Biomedicals

0215013890
ICNA0215013890EA 124 EUR
ICNA0215013890
Mineralöl, Flüssigkeit
Mineralöl

A mineral oil is any of various colorless, odorless, light mixtures of alkanes in the C15 to C40 range from a non-vegetable (mineral) source, particularly a distillate of petroleum. Most often, mineral oil is a liquid by-product of the distillation of petroleum to produce gasoline and other petroleum-based products from crude oil. A mineral oil in this sense is a transparent, colorless oil composed mainly of alkanes and cyclic paraffins, related to petroleum jelly (also known as "white petrolatum").


  • Viscosity: 65.80 - 71.00 Centistokes (at 40 °C) (Lit.)
  • Refractive Index: n20/D 1.467 (lit.)


Mineral oil is used as an overlay covering microdrops of culture medium in petri dishes, during the culture of oocytes and embryos in IVF and related procedures. It also has its applications in food processing, cosmetics and pharmaceuticals.

Siedepunkt: 218…643 °C (1013 hPa)
Dichte: 0,81…0.894 (15 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00131611
CAS-Nummer: 8012-95-1

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
FTIR Conforms to Standard

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR