Adenosin-5'-triphosphat Dinatriumsalz (ATP Dinatriumsalz) Hydrat ≥96%, weißes Pulver

Lieferant: MP Biomedicals

Synonyme: ATP disodium salt hydrate, 5′-ATP-Na2 ATP disodium salt hydrate, Adenosine 5'-Triphosphate Disodium Salt Hydrate, Adenosine-5'-triphosphate disodium salt hydrate, 5'-ATP 2Na Hydrate, ((2R,3S,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl tetrahydrogen triphosphate disodium salt hydrate, ((2R,3S,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl tetrahydrogen triphosphate, disodium salt xhydrate, Adenosine 5'-triphosphoric acid disodium hydrate

0215026680 0215026601 0215026605 0215026610 0215026625
ICNA0215026680EA 936 EUR
ICNA0215026680 ICNA0215026601 ICNA0215026605 ICNA0215026610 ICNA0215026625
Adenosin-5'-triphosphat Dinatriumsalz (ATP Dinatriumsalz) Hydrat ≥96%, weißes Pulver
Adenosin-5'-triphosphat Dinatriumsalz (ATP Dinatriumsalz) Hydrat

Adenosine 5'-triphosphate (ATP) disodium salt hydrate is a multifunctional nucleotide, which is essential in a variety of cellular functions.


Adenosine 5′-triphosphate (ATP) is a central component of energy storage and metabolism in vivo. ATP is used in many cellular processes, respiration, biosynthetic reactions, motility, and cell division. ATP is a substrate of many kinases involved in cell signaling and of adenylate cyclase(s) that produce the second messenger cAMP. ATP provides the metabolic energy to drive metabolic pumps. ATP serves as a coenzyme in a wide array of enzymatic reactions.


P2 purinergic agonist; increases activity of Ca2+-activated K+ channels; substrate for ATP-dependent enzyme systems.


Soluble in aqueous solution (50 mg/ml - clear to slightly hazy solution).

Formel: C₁₀H₁₄N₅Na₂O₁₃P₃·xH₂O
Schmelzpunkt: 185…190 °C (Lit.)
Lagertemperatur: Gefrierschrank
MDL: MFCD00150755
CAS-Nummer: 34369-07-8
EINECS: 213-579-1

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
Purity ≥96%
Moisture Report Result
FTIR Conforms to Standard
UV/Vis: l max (water) 258 ± 2 nm
OD 250/OD 259 0.75 - 0.83
pH (5% aq soln) Acidic to Litmus Paper
Solubility (5% aq soln) Clear to Slightly Hazy Solution


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR