Amberlite® IRA-900 (Cl), Perlen (stark basischer Anionenaustauscher)
Lieferant: MP Biomedicals
Chloride form quaternary ammonium functionality.Strongly basic, macroreticular resin of moderately high porosity with benzyltrialkylammonium functionality.
Amberlite® IRA-900 is a strongly basic (type I) anion exchange resin for decolorizing intermediate to light solutions, concentrating heparin at neutral pH, removing metal ions, etc. Used for recovery of high molecular weight organics from process streams; sugar decolorising.
Practically insoluble in water.
Bitte beachten Research Use Only (RUO).
Spezifikation Testergebnisse
Identity Test | Passes |