1-Aminocyclopropancarbonsäure, weißes Pulver

Lieferant: MP Biomedicals
Achtung

Synonyme: 1-Amino-1-cyclopropanecarboxylic acid

0215034883
ICNA0215034883EA 215 EUR
ICNA0215034883
1-Aminocyclopropancarbonsäure, weißes Pulver
1-Aminocyclopropanecarboxylic acid

1-Aminocyclopropane-1-carboxylic acid is an intermediate in the conversion of methionine to ethylene during the ripening in apples. It also plays an important role in the biosynthesis of the plant hormone ethylene.


Soluble in water (49,00 - 51,00 mg/ml).

Formel: C₄H₇NO₂
Molecular Weight: 101.11 g/mol
Schmelzpunkt: 255…262 °C (Zers., Lit.)
Lagertemperatur: Gefrierschrank
MDL: MFCD00009944
CAS-Nummer: 22059-21-8

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
FTIR Conforms to Standard


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR