Bismarckbraun G, dunkelbraunes Pulver C.I. 21000

Lieferant: MP Biomedicals

0215047325 0215047380
ICNA0215047325EA 37.1 EUR
ICNA0215047325 ICNA0215047380
Bismarckbraun G, dunkelbraunes Pulver C.I. 21000
Bismarckbraun G

Bismarck brown Y is a Plasma stain used for staining mucine, cartilage and goblet cells.


Soluble in water, ethanol, acetic acid, nitric acid; slightly soluble in ethanol and celosolve; practically insoluble in acetone, benzene carbon tetrachloride.

Formel: C₁₈H₁₈N8·2HCl
Lagertemperatur: Raumtemperatur
MDL: MFCD00012977
CAS-Nummer: 10114-58-6
EINECS: 233-314-3
Merck Index: 12,01294

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
UV/Vis: l max 452 - 457 nm (50% EtOH, few drops of 1N HCl)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR