Brillantgrün (Malachitgrün G), grünes Pulver C.I. 42040

Lieferant: MP Biomedicals
Achtung

Synonyme: Ethyl Green, CI 42040, Basic Green 1, N-(4-((4-(Diethylamino)phenyl)(phenyl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-ethylethanaminium hydrogen sulphate, Diamond Green, Malachite Green G, Emerald Green, Astradiamant green GX, Solid Green JO, Brilliant Green hydrogen sulphate

0215051625 0215051680
ICNA0215051625EA 31.9 EUR
ICNA0215051625 ICNA0215051680
Brillantgrün (Malachitgrün G), grünes Pulver C.I. 42040
Brillantgrün (Malachitgrün G)

Brilliant Green is one of the triarylmethane dyes. It is closely related to malachite green.


Soluble in water (1 mg/ml), ethanol, methanol.


Bitte beachten Research Use Only (RUO).

Formel: C₂₇H₃₄N₂O₄S
MDL: MFCD00011880
CAS-Nummer: 633-03-4

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
UV/VIS: λmax (water) 625 ± 2 nm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR