Bromthymolblau Natriumsalz, Pulver

Lieferant: MP Biomedicals

0215052405 0215052425
ICNA0215052405EA 35.9 EUR
ICNA0215052405 ICNA0215052425
Bromthymolblau Natriumsalz, Pulver
Bromthymolblau Natriumsalz
Formel: C₂₇H₂₇Br₂NaO₅S
MDL: MFCD00077263
CAS-Nummer: 34722-90-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR