Cäsiumchlorid 99,999, powder_white, ultrapure
Lieferant: MP Biomedicals
Synonyme:
Cesium Chloride
0215058905
0215058925
0215058980
0215058990
0215058991
ICNA0215058905EA
63.2
EUR
ICNA0215058905
ICNA0215058925
ICNA0215058980
ICNA0215058990
ICNA0215058991
Cäsiumchlorid 99,999, powder_white, ultrapure
Cesium chloride
Cesium chloride is used for the preparation of electrically conducting glasses. It is also used to make solutions for the separation of RNA from DNA by density gradient centrifugation.
- A260 of 50% solution < 0.02
- Solutions are clear and colorless
- Especially suited for critical density gradient techniques.
pH: 5.5 - 6.5 (1% aq soln)
Very soluble in water. Soluble in ethanol.
Formel:
CsCl Molecular Weight: 168.36 g/mol Siedepunkt: 1303 Schmelzpunkt: 646 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010955 CAS-Nummer: 7647-17-8 Merck Index: 13,02021 |
Spezifikation Testergebnisse
Purity | 99.999% |
pH | 5,5 - 6,5 (1% aq soln) |
Refractive Index | 1,6418 (Lit.) |
Barium | ≤1 ppm |
Magnesium | ≤0,5 ppm |
Potassium | ≤2 ppm |
Iron | ≤1 ppm |
Calcium | ≤1 ppm |
OD260 nm (50% aq soln) | <0.02 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...