Dabsylchlorid, dunkelrotes Pulver

Lieferant: MP Biomedicals
Gefahr

Synonyme: DABS-Cl, 4-Dimethylaminoazobenzene-4'-sulphonyl Chloride, 4-((4-(Dimethylamino)phenyl)diazenyl)benzenesulphonyl chloride

0215093183
ICNA0215093183EA 62.1 EUR
ICNA0215093183
Dabsylchlorid, dunkelrotes Pulver
Dabsyl chloride

4-Dimethylaminoazobenzene-4'-sulfonyl Chloride (dabsyl chloride) is a chromophoric label for amino acids, peptides and proteins.


For the qualitative and quantitative identification of amino acids and for the determination of N-terminal amino acids. Used in the determination of primary and secondary amines by HPLC.


Soluble in acetonitrile, DMF; slightly soluble in acetone.


Bitte beachten Research Use Only (RUO).

Formel: C₁₄H₁₄ClN₃O₂S
Molecular Weight: 323.80 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00007444
CAS-Nummer: 56512-49-3

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
FTIR Conforms to Standard


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR