L(-)-Glutathion (oxidierte Form), kristallines Pulver
Lieferant: MP Biomedicals
Synonyme:
GSSG, Bi(glutathion-S-yl), L(-)-Glutathion, oxidiert
Amino Acid Sequence:Glu-Cys-Gly, Glu-Cys-Gly [disulfide bridge: 2a-2b].
In vivo, Glutathione disulfide (GSSG) is reduced back to glutathione (GSH) via the NADPH-dependent enzyme glutathione reductase. The ratio of GSH to GSSG is often used to gauge the level of oxidative stress in cells, with higher concentrations of GSSG implying more oxidative stress
Soluble in water (3,3%): clear, colorless solution.
Bitte beachten Research Use Only (RUO).
Formel:
C₂₀H₃₂N₆O₁₂S₂ Molecular Weight: 612.64 g/mol Schmelzpunkt: 185 Lagertemperatur: Kühlschrank |
MDL:
MFCD00063106 CAS-Nummer: 27025-41-8 |
Spezifikation Testergebnisse
Identity Test | Passes |
pH (3.3% aq soln) | Acidic to Litmus Paper |
Solubility (3.3% aq soln) | Clear, Colorless Solution |
Optical Rotation (c=1, water) | –85 ° to –110 ° |
FTIR | Conforms to Standard |
Moisture | For Information Only |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...