Glycerin, Flüssigkeit
Lieferant: MP Biomedicals
Synonyme:
1,2,3-Propantriol, Glycerol
0215119490
0215119491
ICNA0215119490EA
53.3
EUR
ICNA0215119490
ICNA0215119491
Glycerin, Flüssigkeit
Glycerin
Formel:
HOCH₂CH(OH)CH₂OH |
MDL:
MFCD00004722 CAS-Nummer: 56-81-5 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...