Naphthalin ≥99%, weißes kristallines Pulver für die Szintillation

Lieferant: MP Biomedicals
Achtung

0215172680
ICNA0215172680EA 42.9 EUR
ICNA0215172680
Naphthalin ≥99%, weißes kristallines Pulver für die Szintillation
Naphthalene

Soluble in hydronaphthalenes, ether, carbon disulfide, chloroform, carbon tetrachloride, benzene, toluene, ethanol, methanol (100 mg/ml - clear, colorless solution); insoluble in water.


Naphthalene was used in liquid-phase exfoliation of graphite in organic solvents for the production of graphene sheets.
It was used in preparation of carbon-coated Si70Sn30 nanoparticles
It was used as fluorescent probe to study the aggregation behavior of sodium cholate
It was used to investigate influence of added short chain linear and branched alcohols on the binding of 1:1 complex of naphthalene and β-cyclodextrin


Soluble in hydronaphthalenes, ether, carbon disulfide, chloroform, carbon tetrachloride, benzene, toluene, ethanol, methanol (100 mg/ml - clear, colorless solution); insoluble in water.

Formel: C₁₀H₈
Molecular Weight: 128,17 g/mol
MDL: MFCD00001742
CAS-Nummer: 91-20-3
UN: 1334
ADR: 4.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c87c606cb828e0f39bab8b9d50c2c26501e1637d&vid=9e4bc21353014391fc8486208c8757e6375e2ef5&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
FTIR Conforms to Standard
Melting Point 78 - 83 °C
Purity ≥99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR