Ölsäure 99, Liquid_clear, colorless

Lieferant: MP Biomedicals

Synonyme: cis-9-Octadecensäure

0215178101 0215178105 0215178125
ICNA0215178101EA 68.6 EUR
ICNA0215178101 ICNA0215178105 ICNA0215178125
Ölsäure 99, Liquid_clear, colorless
Ölsäure

Oleic acid is a mono-unsaturated omega-9 fatty acid created by dehydrogenation of stearic acid. It is then further dehydrogenated and elongated to create members of the n-9 family of fatty acids.


Oleic acid is used as emollient. Small amounts of oleic acid are used as an excipient in pharmaceuticals. It is used as an emulsifying or solubilizing agent in aerosol products. Oleic acid have been shown to be important for the growth and productivity of Chinese Hamster Ovary (CHO) cells. Functions of Oleic Acid in Cell Culture Systems: Long-term energy storage: energy derived from NADPH and ATP is stored in fatty acids. Fatty acids are esterified to a glycerol backbone to form a group of compounds known as mono-, di- and tri- glycerides (neutral fats). Energy is released when fatty acids are degraded.


Soluble in methanol (40 mg/ml - clear solution), chloroform, benzene or fixed and volatile oils. Insoluble in water.


Bitte beachten Research Use Only (RUO).

Formel: C₁₈H₃₄O₂
Molecular Weight: 282.47 g/mol
Siedepunkt: 286
Schmelzpunkt: 13…14 °C (Lit.)
Flammpunkt: 180
Lagertemperatur: Gefrierschrank
MDL: MFCD00064242
CAS-Nummer: 112-80-1
Merck Index: 13,06898
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e5aecc5dce4d0318386a45d7adb7c5822631010a&vid=892a2df4ecab3679b06983b84cd27ad334ef961f&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
Purity ≥99%
FTIR Conforms to Standard


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR