Phosphoenol pyruvic acid trisodium salt hydrate

Lieferant: MP Biomedicals

15187291 0215187225 0215187280 0215187283 0215187290 0215187294
ICNA15187291EA 437 EUR
ICNA15187291 ICNA0215187225 ICNA0215187280 ICNA0215187283 ICNA0215187290 ICNA0215187294
Phosphoenol pyruvic acid trisodium salt hydrate
Phosphoenol pyruvic acid trisodium salt hydrate

Soluble in water.

Formel: C₃H₂Na₃O₆P
Molecular Weight: 233.986 g/mol
MDL: MFCD00150737
CAS-Nummer: 5541-93-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR