di-Kaliumhydrogenphosphat Trihydrat ≥99%, Kristalle

Lieferant: MP Biomedicals

Synonyme: di-Kaliumphosphat Trihydrat, Dikaliumhydrogenphosphat Trihydrat, sek. Kaliumphosphat Trihydrat

02151946.1 02151946.5 0215194610 0215194601
ICNA02151946.1EA 38.5 EUR
ICNA02151946.1 ICNA02151946.5 ICNA0215194610 ICNA0215194601
di-Kaliumhydrogenphosphat Trihydrat ≥99%, Kristalle
di-Kaliumhydrogenphosphat Trihydrat

Soluble in water (100 mg/ml) ; Insoluble to slightly soluble in ethanol; insoluble in alcohol.


For phosphate buffers, the pH increases with decreasing temperature. Compared with a buffer at 25 °C, buffers at 4 °C will be 0,08 pH unit higher and a buffer at 37 °C will be 0,025 pH unit lower. The concentration of phosphate also influences pH. ; Typical Buffer Preparation/Formulations: 1 M Potassium Phosphate Monobasic, 1 M Potassium Phosphate Dibasic, 5 M Sodium Chloride

Formel: K₂HPO₄·3H₂O
Molecular Weight: 228.22 g/mol
Dichte: 2.3 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00149926
CAS-Nummer: 16788-57-1
EINECS: 231-834-5

Bestellen

Spezifikation Testergebnisse

Form Clear Granular Crystals
Storage Store at room temperature (15 to 30 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR