Propylenglycol, Flüssigkeit

Lieferant: MP Biomedicals

Synonyme: Propylenglycol, (±)-1,2-Propylenglycol, (±)-1,2-Dihydroxypropan

0215195790 0215195791
ICNA0215195790EA 27 EUR
ICNA0215195790 ICNA0215195791
Propylenglycol, Flüssigkeit
Propylenglycol

Propylene glycol is a clear, colorless and hygroscopic liquid. Propylene glycol contains an asymmetrical carbon atom, so it exists in two stereoisomers.


The freezing point of water is depressed when mixed with propylene glycol due to increased opportunity for hydrogen bonding.
Propylene glycol is a solvent in many pharmaceuticals, including oral, injectable and topical formulations, such as for diazepam and lorazepam that are insoluble in water, use propylene glycol as a solvent in their clinical, injectable forms. It is also used as automotive antifreeze.

Formel: CH₃CH(OH)CH₂OH
Molecular Weight: 76.1 g/mol
Siedepunkt: 187.9 °C (1013 hPa) (Lit.)
Schmelzpunkt: –60 °C (Lit.)
Dichte: 1.04 g/cm³ (20 °C) (Lit.)
Flammpunkt: 99
Lagertemperatur: Raumtemperatur
MDL: MFCD00064272
CAS-Nummer: 57-55-6
Merck Index: 13,07947
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f2af195b87767feb8d1baca34925153d6f97ae2d&vid=a598a9042847dc3ce52daed1a9326b592b163170&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
FTIR Conforms to Standard


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR