Reactive blue 4, Pulver

Lieferant: MP Biomedicals

0215201610
ICNA0215201610EA 25.2 EUR
ICNA0215201610
Reactive blue 4, Pulver
Reactive blue 4
Formel: C₂₃H₁₄Cl₂N₆O₈S₂
MDL: MFCD00001218
CAS-Nummer: 13324-20-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR