Reactive red 4, Pulver C.I. 18105

Lieferant: MP Biomedicals

Synonyme: tetrapotassium 5-benzamido-3-{2-[5-({4-chloro-6-[(4-sulphonatophenyl)amino]-1,3,5-triazin-2-yl}amino)-2-sulphonatophenyl]diazen-1-yl}-4-hydroxynaphthalene-2,7-disulphonate, Procion Reactive Red 4, Cibacron Brilliant Red 3B-A

0215201705 0215201725
ICNA0215201705EA 73 EUR
ICNA0215201705 ICNA0215201725
Reactive red 4, Pulver C.I. 18105
Reactive red 4

A monochlorotriazine dye that inhibits phosphotransferase acitvity


Soluble in water. Very slightly soluble in ethanol and methanol.

Formel: C₃₂H₁₉ClN₈O₁₄S₄Na₄
Molecular Weight: 995.2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00009690
CAS-Nummer: 17681-50-4

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
λmax (ethanol) 282 ±2 nm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR