Spectinomycin dihydrochloride pentahydrate
Lieferant: MP Biomedicals
Synonyme:
(2R,4aR,5aR,6S,7S,8R,9S,9aR,10aS)-4a,7,9-Trihydroxy-2-methyl-6,8-bis(methylamino)decahydro-4H-benzo[b]pyrano[2,3-e][1,4]dioxin-4-one dihydrochloride pentahydrate
0215206701
0215206725
ICNA0215206701EA
32.9
EUR
ICNA0215206701
ICNA0215206725
Spectinomycin dihydrochloride pentahydrate
Spectinomycin dihydrochloride pentahydrate
Soluble in water, methanol, propyleneglycol; very soluble benzene, chloroform, ethanol and acetone.
Formel:
C₁₄H₂₄N₂O₇·2HCl·5H₂O |
MDL:
MFCD00150886 CAS-Nummer: 22189-32-8 |
Spezifikation Testergebnisse
Synonyms | U18409AE; Actinospectacin; M141 |
Form | White Powder |
Storage | Store at +4 °C |
Heavy Metals | ≤0,005% |
Loss on Drying |
|
Optical Rotation | Positive |
Potency | ≥500 μg/mg |
Source | Streptomyces sp. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...