Spermin (N,N'-Bis(3-aminopropyl)-1,4-butandiamin) ≥97%, farbloser Feststoff

Lieferant: MP Biomedicals
Gefahr

Synonyme: N1,N1'-(Butane-1,4-diyl)bis(propane-1,3-diamine), Spermine

02152070.5 0215207001 0215207005
ICNA02152070.5EA 51.9 EUR
ICNA02152070.5 ICNA0215207001 ICNA0215207005
Spermin (N,N'-Bis(3-aminopropyl)-1,4-butandiamin) ≥97%, farbloser Feststoff
Spermin (N,N'-Bis(3-aminopropyl)-1,4-butandiamin)

Spermine is a polyamine involved in cellular metabolism found in all eukaryotic cells. Formed from spermidine through the addition of aminopropyl group to spermidine by spermine synthase. It is found as a polycation at physiological pH.


Mixed NMDA glutamate receptor agonist/antagonist at the polyamine site. Neuroprotective effects have been observed at high concentrations (1 mM), while neurotoxicity is observed at lower concentrations. It enhances agonist effectiveness at the strychnine-insensitive glycine site. Plays a role in cellular proliferation and differentiation; inhibits neuronal nitric oxide synthase (nNOS).


Soluble in water (1% w/v), lower alcohol, chloroform; practically insoluble in ether, benzene, petroleum ether.


Bitte beachten Research Use Only (RUO). Desiccate, store under nitrogen.

Formel: C₁₀H₂₆N₄
Siedepunkt: 141…142 °C (0,5 mmHg)
Schmelzpunkt: 28…30 °C (Lit.)
Lagertemperatur: Kühlschrank
MDL: MFCD00008215
CAS-Nummer: 71-44-3

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
Purity ≥97%
FTIR Conforms to Standard
pH (0.5% aq soln) 11 - 14


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR