Tween® 85 (Polysorbat)

Lieferant: MP Biomedicals

Synonyme: Polyoxyethylenesorbitan Trioleate

0215248780 0215248790
ICNA0215248780EA 29.6 EUR
ICNA0215248780 ICNA0215248790
Tween® 85 (Polysorbat)
Tween® 85 (Polysorbat)
Dichte: 1.02 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD01779641
CAS-Nummer: 9005-70-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR